Learn More
Bel-Art™ Pipet Vacuum Aspirator Collection System
Shop All SP Scienceware Products
Click to view available options
Type:
Pipet Vacuum Aspirator Collection System
Replacement Hydrophobic Filter
Unit Size:
Each
Pack of 12
Description
- Built-in pump quickly aspirates fluids from pipet tip, Pasteur pipet or serological pipet inserted into the collection tube
- Maintains low noise level (76 db at 1 meter) and minimal vibration
- Excellent for fluid collection from tissue and/or cell culture, Petri dishes, plates, flasks and tubes
- Chemical and corrosion resistant polypropylene collection bottle holds 3800 mL (1 gallon) fluid wastes and has a safety release vent to prevent vacuum implosion
- A replaceable hydrophobic filter seals itself if it becomes wet protecting the pump motor from fluid contamination
- Lightweight and compact design will not crowd workspace in a hood or at the bench
Specifications
Specifications
| For Use With | Pipettes |
| Material | Plastic |
| Type | Pipet Vacuum Aspirator Collection System |
| Volume (Metric) | 3800 mL |
| For Use With (Equipment) | Media Bottles |