Learn More
BD Rat TNF ELISA Set
Design an immunoassay with different sensitivity and dynamic range
Brand: BD 558535
Additional Details : Weight : 0.04000kg
Includes: Capture Antibody: Anti-rat TNF-α, Detection Antibody: Biotinylated anti-rat TNF-α, Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate (SAv-HRP) (250X), Standard: Recombinant rat TNF-α (lyophilized)
Description
- Flexible assay
- Calibrated against purified Baculovirus-expressed recombinant rat TNF
ELISA
Specifications
31.3 to 2000pg/mL | |
20 96-well Plates | |
Biotin anti-Rat TNF (250X) (detection) Antibody, Instruction/analysis certificate, Lyophilized recombinant Rat TNF (standard), Purified anti-Rat TNF (250X) (capture) antibody, Streptavidin-HRP Conjugate (250X) | |
2°C to 8°C |
Rat TNF ELISA set | |
To develop ELISA for natural or recombinant Rat TNF in Serum, Plasma and cell culture supernatants | |
20 Plates |
Research use only.