Learn More
BD Rat IL-10 ELISA Set
To develop ELISA for natural or recombinant Rat IL-10 in Serum, Plasma and cell culture supernatants
Brand: BD 555134
Additional Details : Weight : 0.16000kg
Description
The OptEIA™ set contains components necessary to develop enzyme-linked immunosorbent assays (ELISA) for natural or recombinant rat IL-10 in serum, plasma, and cell culture supernatants.
- Capture Antibody: anti-rat IL-10 monoclonal antibody
- Detection Antibody: biotinylated anti-rat IL-10 monoclonal antibody
- Enzyme Reagent: streptavidin-horseradish peroxidase conjugate (SAv-HRP)
- Standards: recombinant rat IL-10, lyophilized
Specifications
Rat IL-10 ELISA set | |
To develop ELISA for natural or recombinant Rat IL-10 in Serum, Plasma and cell culture supernatants | |
20 Plates |
20 96-well Plates | |
anti-Rat IL-10 (capture) antibody, Biotinylated anti-Rat IL-10 (detection) antibody, Instruction/analysis certificate, Lyophilized recombinant Rat IL-10 (standard), Streptavidin-HRP | |
2°C to 8°C |