Learn More
BD Mouse Intracellular Cytokine Staining Starter Kit
Serves as an introduction to the intracellular staining technique
Brand:  BD 559311
Additional Details : Weight : 2.00000kg
Product Code. 15839698
  • £1185.00 / 1 set
Estimated Shipment: 30-05-2024
to see stock.
Add to basket
Includes: BD PharMingen Stain Buffer (FBS); Cytofix/Cytoperm Buffer; Perm/Wash Buffer; PE-conjugated anti-human or mouse IL-2; PE-conjugated anti-human or mouse IFN-g; PE-conjugated anti-human or mouse TNF-a; PE-conjugated isotype control cocktail; Purified blocking antibody cocktail; Leukocyte Activation Cocktail; MiCK-1 or HiCK-1 Cytokine Positive Control Cells; Staining Kit Manual
Description
Description
  • Contains the essential reagents for the activation of a mouse spleen cell population and their subsequent intracellular staining to detect IL-2, IFN-γ-, and TNF-a-producing cells
Specifications
Specifications
IL-2, IFN-g- and TNF-a
25 Tests
4°C