missing translation for 'onlineSavingsMsg'
Learn More

BD Mouse intracellular CytoKine-2 Positive Control Cells, MiCK-2

Contains fixed,non-permeabilized mouse lymphoid cells

Brand:  BD 554653

Additional Details : Weight : 0.01000kg

Product Code. 15897818

  • £2165.00 / Each
Estimated Shipment: 30-05-2024
to see stock.
Add to basket

Description

Description

  • Cells easily express detectable levels of intracellular IL-4, IL-10 or GM-CSF
  • Prepared by stimulating mouse spleen cells in the presence of a protein transport inhibitor
  • Contains measurable proportions of cytokine- producing cells
  • Storage buffer: frozen in FBS and 10% DMSO

Intracellular staining (flow cytometry)

Specifications

Specifications

Frozen
5 x 106 cells/mL
Intracellular staining
1 mL
Light Brown
IL-4, IL-10 or GM-CSF
≈5 x 106 Activated Mouse splenocytes (20 test samples at 2.5 x 105 cells/test) fixed and suspended in 1mL
-80°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For research use only.