missing translation for 'onlineSavingsMsg'
Learn More

A&D Weighing™ BA-125DTE Premium-Level Micro/Semi-Micro Balance

The BA and BA-T series bring maximum operability, enhanced functionality and a broader range of solutions. All models come standard with an external ionizer for quick and easy elimination of static.

Brand:  A&D Weighing™ BA-125DTE

Additional Details : Weight : 6.60000kg

Product Code. 17877590

  • £4770.00 / Each
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Multi-functional color touchscreen displays for intuitive operation of advanced features
  • Automatic and programmable opening/closing of the breeze break doors using non-contact IR sensors
  • Detachable breeze break unit with easily clean separable glass panes for better cleanability
  • No grooves for the sliding doors so that contaminants do not accumulate
  • Additional inner breeze break for 0.001 mg models to increase stability
  • External ionizer for instant static removal
  • User access control (UAC) with user rights management and password lock functions
  • History information for log-in/log-out, operation, sensitivity adjustment, and impact shock detection (ISD) Impact shock detection (ISD) providing feedback to prevent damage to the weight sensor (patented)
  • Formulation mode for quick, accurate weighing of multiple samples to mix
  • HPLC mode for preparing buffer solutions for high-performance liquid chromatography
  • Daily/periodic balance check support
  • Smart routine check (SRC) function
  • On-screen leveling assist
  • 'AD-Just' automatic self-sensitivity adjustment using the internal weight Internal repeatability test to quickly assess the performance under a given environment
  • Data memory function
  • Selectable response characteristics for trading faster weighing speed for higher display stability or vice versa GLP/GMP/GCP/ISO compliant output to meet documentation requirements
  • Universal Flexi Coms (UFC) to customize printout content and layout
  • Gross/Net/Tare output Counting mode with the Automatic Counting Accuracy Improvement (ACAI) function
  • Percent mode function for target weighing or checking sample variation
  • Statistical calculation function (SCF) to facilitate the analysis of measurements
  • Auto power ON and OFF functions Underhook for weighing a magnetic material as well as measuring density
Specifications

Specifications

RS-232
CE, UKCA, GLP/GMP
51 g/120 g
0.01 mg/0.1 mg
Display Cover, AC Adapter, 1.8 m USB Cable, Tweezers for Calibration Weight, Microspatula
Manual with On-screen Leveling Assist
Aluminum, Plastic, Glass
7 sec.
5°C to 40°C
3 Years
6.8 in.
11.2 in.
284 mm
17.7 in.
3.4 in.
14.77 lb.
100/240 V
Internal
RS-232C (D-Sub 9P), USB-A, USB-B (Mini B), Ethernet (TCP/IP), External Key Input (2)
Color Touchscreen
≤85% at 5°C to 40°C
English, French, German, Italian, Dutch, Spanish, Portuguese, Russian, Korean, Chinese, Japanese
±0.2 mg
0.03 mg/0.1 mg
41°F to 104°F
Analytical Balance
Weigh, Count, Percent, Formulation, HPLC
173 mm
6.5 in.
166 mm
450 mm
85 mm
6.7 kg
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers