Learn More
Axygen™ Tip Refill System, Empty Racks
Easy-to-load refill racks for Axygen Tip Refill System. For use with P2/P10 Gilson-Style and 200μL Universal fit inserts.
Brand: Axygen™ RFL-R-1200
Code : A0
Additional Details : Weight : 0.99000kg
Description
Axygen™ Tip Refill System contains durable refill rack that can be easily refilled by hand and is autoclavable. These translucent Racks includes color-coded inserts for tip identification and are certified RNase-/DNase free. They are available in different colors and are packaged in bulk or racks.Specifications
Empty Refill Rack/Lid | |
Non-sterile | |
RFL-R-1200 |
Clear | |
P2/P10 Gilson-Style and 200μL Universal fit inserts | |
10 Racks/Pack; 3 Packs/Case |