Learn More
Excel Healthcare Ltd™ Autoclave Printer Rolls
Thermo printer rolls
Brand: Excel Healthcare Ltd™ 299
Additional Details : Weight : 1.00000kg
Description
- Excel Healthcare Has A Range Of Testing Products That Are Compatible With Our Range Of Steam Sterilisers
- Manual Testing Of Steam Sterilisers Is Now Considered An Essential Requirement And Must Be Performed By The Operator On A Daily Basis
- Htm 01-05 States That: Daily Tests Must Be Performed By The Operator Or User And If The Steriliser Fails To Meet Any Of The Test Requirements The Steriliser Must Be Withdrawn From Service And Advice Sought From The Manufacturer Or Service Provider
- The Daily Tests Consist Of Steam Penetration Test (Vacuum Sterilisers Only) And Automatic Control Test. These Tests Should Be Dated, Signed And Recorded In The Steriliser Log Book
- These Tests Are Easy To Complete And Record, Giving Full Performance Traceability Of Your Vacuum Steriliser
Specifications
57 mm | |
Printer Roll (single) |