missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Arium™ Smart Station PW
Arium™ Smart Station flexible remote dispenser for pure water
£1740.00 - £1745.00
Specifications
Capacity (Metric) | 50 mL to 50 L |
---|---|
Feedwater Source | Pure |
Product Water Type | Pure Water |
Display | Touch display |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
17264233
|
Sartorius
H2O-ARST-P-T |
Smart Station Benchtop |
£1745.00
1 unit |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17254233
|
Sartorius
H2O-ARST-P-B |
Smart Station Wall-Mounted |
£1740.00
1 unit |
Estimated Shipment: 23-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Height adjustable display/dispense station for dispensing pure water
- Compact design for easy and space-saving integration into lab environment
- Intuitive touch-activated color display with direct access to important functions
- Flexible and stepless height adjustment to fill different sized containers and beakers
- Ergonomic design can be optimized for either right or left hand operation
- Accurate volume dispense for fast and precise buffer and sample prep
- Adjustable, splash-free flow rate from drop-by-drop to 2 L/min
Equipment supplied:
- Display/dispense unit with wall bracket
- Power supply including:
- IEC connector
- Connection set
For intended use with:
- Arium™ Bagtank
- Arium™ Pro
- Arium™ Comfort
Specifications
50 mL to 50 L | |
Pure | |
Pure Water | |
Touch display |