missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Arium™ Mini Essential for Production of ASTM Type 1 Ultrapure Water with UV
Compact Arium™ mini laboratory water systems have been designed for Type 1 ultrapure water requirements of 10 liters per day and are ideal for use in preparation of buffers, media and samples, both in life sciences applications and in analytical laboratory procedures.
Brand: Sartorius H2O-MU-UV-T
Description
- Reliable: Delivers consistently high water quality for reliable and reproducible results
- Intuitive: Touch-activated color display with direct access to all important dispensing options
- Innovative: saves time-intensive tank cleaning
- Compact: Space-saving, with a width of only 28 cm, UV Lamp for Critical Analyses (185 | 254 nm)
- Optimal temperature gradient due to horizontal installation
- Degradation of total organic carbons (TOCs) by irradiation (< 5 ppb)
- Inactivates microorganisms by damaging their DNA
Specifications
EU Declaration of Conformity, CSA Declaration of Conformity, QA Certificate | |
10 L | |
Pretreated | |
Type 1 | |
< 5 ppb | |
Lab Water System with UV Lamp | |
0 to 6.9 bar; (0 to approx. 100 psi); recommended > 2 bar (>29 psi) | |
20.8 x 11 x 20 in. | |
509 mm | |
100/240 V |
< 100 μS/cm, compensated to 25°C | |
Water pretreated by reverse osmosis, distillation or deionization | |
Up to 1.0 L/min. | |
18.2 Mohm.cm at 25°C | |
<10 ppb | |
Touch activated | |
Arium mini accessories | |
280 mm | |
100/240 VAC 50/60 Hz 2.0 A max | |
Input supply voltage VDC +24 (±10%) Current consumption, max. A 3.0 |