missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PITPNM3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521903
This item is not returnable.
View return policy
Description
Recommended positive controls: MCF-7, MDA-MB-231. Predicted reactivity: Mouse (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of a family of membrane-associated phosphatidylinositol transfer domain-containing proteins. The calcium-binding protein has phosphatidylinositol transfer activity and interacts with the protein tyrosine kinase PTK2B. The protein is homologous to a Drosophila protein that is implicated in the visual transduction pathway in flies. Mutations in this gene result in autosomal dominant cone dystrophy. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| PITPNM3 | |
| Polyclonal | |
| Unconjugated | |
| PITPNM3 | |
| A330068P14Rik; ACKR6; AI848332; atypical chemokine receptor 6; cone rod dystrophy 5; CORD5; Gm880; Membrane-associated phosphatidylinositol transfer protein 3; MGC157740; MGC157741; NIR1; NIR-1; phosphatidylinositol transfer protein, membrane-associated 3; PITPnm 3; PITPNM family member 3; Pitpnm3; Pyk2 N-terminal domain-interacting receptor 1; RDGBA3; retinal degeneration B alpha 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83394 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q9BZ71 | |
| PITPNM3 | |
| Synthetic peptide corresponding to a region within amino acids 912 and 974 of NIR1 (Uniprot ID#Q9BZ71) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction