missing translation for 'onlineSavingsMsg'
Learn More

Ammonia Solution, 35%, Extra Pure, SLR, d=0.88, Fisher Chemical™

Catalog Number p-7072798
29.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


28.17 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.


Click to view available options
Quantity:
1 L
2.5 L
1336-21-6
H5NO
11418
1336-21-6
H5NO
11418

H5NO, CAS Number-1336-21-6, 2.5L, Colorless, 35.05g/mol, 11418, Plastic Coated Glass bottle, 12, Liquid

TRUSTED_SUSTAINABILITY
pH 12
Color Colorless
Quantity 1 L
Formula Weight 35.05g/mol
Packaging Plastic Coated Glass bottle
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonia solution 35%
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Ammonium hydroxide,Ammonia
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'