missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonia Solution, 35%, Certified AR for Analysis, d=0.88, Fisher Chemical™
27.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
HDPE plastic bottle
Plastic Coated Glass bottle
Plastic container
Description
H5NO, CAS Number-1336-21-6, aquammonia, ammonium hydroxide solution, ammonium hydroxide nh4 oh, ammonia water, aqua ammonia, ammonia, monohydrate, ammonium hydroxide, ammonia aqueous, household ammonia, ammonia, aqua, 2.5L, CHEBI:18219, Colorless, 35.05g/mol, VHUUQVKOLVNVRT-UHFFFAOYSA-N
Specifications
pH | 12 |
Color | Colorless |
Quantity | 1 L |
IUPAC Name | azanium;hydroxide |
Formula Weight | 35.05g/mol |
Packaging | Plastic Coated Glass bottle |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'