missing translation for 'onlineSavingsMsg'
Learn More

BUSE Scientific Ammonia Gas

Brand:  BUSE Scientific LBAMM38

Code : 22

Additional Details : CAS Number : 7664-41-7 Weight : 1.50000kg Transport : UN number : 1005 Chem class : 2.3

Product Code. 12776177

  • POA / 170g
Estimated Shipment: 04-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

Specifications

Specifications

170 g
Ammonia
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers