missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Protran™ Supported NC Nitrocellulose Membranes: Sheets

Provides exceptional handling characteristics, allowing reprobing

Brand:  Cytiva 10600053

Code : GH

Additional Details : CAS Number : 9004-70-0 Weight : 0.05500kg


 View more versions of this product

Product Code. 15259834

  • £351.00 / 25 sheets
Estimated Shipment: 19-06-2024
to see stock.
Add to basket

Description

Description

Individual sheets.

  • Membranes consist of nitrocellulose supported by an inert polyester nonwoven material within the membrane
  • The support does not affect transfer conditions or results

Specifications

Specifications

20 cm
25/Pk.
Nitrocellulose, Inert Polyester
20 cm
Sheet
0.2 μm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • amersham protran supported 0.2µm NC 200mm x 200mm

Signal Word
  • Warning

Hazard Category
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose

SDS
Documents

Documents

Certificates
Special Offers

Special Offers