missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Sheets

Use for high sensitivity, resolution, and low background for all labeling and detection systems, including chemiluminescent, fluorescenct, colorimetric and radioactive. Cytiva Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.

Brand:  Cytiva 10600080

Code : GH

Additional Details : CAS Number : 9004-70-0 Weight : 0.01400kg


 View more versions of this product

Product Code. 15259854

  • £172.00 / 25 sheets
Estimated Shipment: 02-07-2024
to see stock.
Add to basket

Description

Description

Use for high sensitivity, resolution, and low background for all labeling and detection systems, including chemiluminescent, fluorescenct, colorimetric and radioactive. Cytiva Amersham™ Protran™ Premium NC Nitrocellulose Membranes: Sheets offer excellent sensitivity, resolution, and low background.

Individual sheets.

 

  • Low background enables a dynamic range for complicated samples
  • Suitable for Western blots, dot-blot assays, and other protein or nucleic acid methods
  • 100% pure nitrocellulose
  • High affinity for proteins
  • Blocking ability

 

Specifications

Specifications

10 cm
Western blots, dot-blot assays, and other protein or nucleic acid methods
Sheet
10 cm
25/Pk.
0.2 μm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Amersham Protran Premium

Signal Word
  • Warning

Hazard Category
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers