missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specac™ Agate Pestle and Mortar 4cm Diameter Bowl
Brand: Specac™ GS03600
Code : Z3
Additional Details : Weight : 0.12000kg
Description
Agate pestle and mortar designed for preparing and grinding samples. The mortar bowl has a 4cm diameter.Specifications
4 cm |