Learn More
Eppendorf™ Adapter for S-4xUniversal Rotor
The Adapter for S-4xUniversal Rotor facilitates the use of specific sizes, shapes, and numbers of vessels in rotor buckets for the S-4xUniversal Rotor.
Brand:  Eppendorf™ 5910757000
Additional Details : Weight : 0.49500kg
 View more versions of this product
Product Code. 15870927
  • £301.00 / Pack of 2
In Stock
to see stock.
Description
Description
  • Suitable for S-4xUniversal rotor buckets
  • Accommodates selected array of tubes, bottles, or plates
Specifications
Specifications
Flat Bottom
102 mm
S-4xUniversal Rotor
2
1 x 750 mL Flat-bottom Bottle
139 mm
1