Learn More
Thermo Scientific™ Heraeus™ Centri-Lab Type D Adapter
Type D adapter for use with centrifuges.
Brand: Thermo Scientific™ 75006455
Code : 51
Additional Details : Weight : 0.20000kg
Description
Type D adapter for use with centrifuges.
Heraeus™ Centri-Lab Type D adapters are suitable for Heraeus™ centrifuge.
- For flat tubes with round shape
Specifications
Tube Adapter | |
15 mL | |
Red |
Borosilicate Glass | |
Heraeus™ Centrifuge | |
16 |