Learn More
Thermo Scientific™ Filtration Hardware, Accessories, and Spare Parts
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.
Brand:  Thermo Scientific™ 710300-0003
Code : A0
Additional Details : Weight : 0.03100kg
 View more versions of this product
Product Code. 10566551
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.
Specifications
Specifications
Clear
13.3 cm2
Sterilization Plates and O-Rings
Nalgene Reusable Filter Holders with Receiver
Cold Sterilization and Clarification of Culture Media, Reagents or Solvents