missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-3,3,3-trifluoro-1-propene, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
It is used as intermediate in production of organ fluorine compounds & as a monomer in fluoropolymer production. It is used as a trifluoromethylated synthetic building block for the synthesis of trifluoromethyl-substituted molecules.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as intermediate in production of organ fluorine compounds & as a monomer in fluoropolymer production. It is used as a trifluoromethylated synthetic building block for the synthesis of trifluoromethyl-substituted molecules.
Solubility
Soluble in most organic solvents, immiscible with water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
It is used as intermediate in production of organ fluorine compounds & as a monomer in fluoropolymer production. It is used as a trifluoromethylated synthetic building block for the synthesis of trifluoromethyl-substituted molecules.
Solubility
Soluble in most organic solvents, immiscible with water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Specifications
Refractive Index | 1.353 |
Quantity | 25 g |
Solubility Information | Soluble in most organic solvents,immiscible with water. |
Formula Weight | 174.95 |
Percent Purity | 97% |
Chemical Name or Material | 2-Bromo-3,3,3-trifluoro-1-propene |
Safety and Handling
Signal Word
- Warning
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H361-Suspected of damaging fertility or the unborn child.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'