missing translation for 'onlineSavingsMsg'
Learn More

2,4,4-Trimethyl-1-pentene, 99%

Product Code. p-3737689 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 10426780

Brand: Thermo Scientific Acros 140181000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-39-1 | C8H16 | 112.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-39-1
Molecular Formula C8H16
Molecular Weight (g/mol) 112.21
MDL Number MFCD00008855
InChI Key FXNDIJDIPNCZQJ-UHFFFAOYSA-N
Synonym 2,4,4-trimethyl-1-pentene, diisobutylene, pentene, 2,4,4-trimethyl, 2,4,4-trimethylpentene, 1-pentene, 2,4,4-trimethyl, 2,2,4-trimethyl-4-pentene, unii-n69l73advf, 1-methyl-1-neopentylethylene, ccris 9103, 2,4,4-trimethylpentene-1
PubChem CID 7868
IUPAC Name 2,4,4-trimethylpent-1-ene
SMILES CC(=C)CC(C)(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -101°C
Color Colorless
Density 0.7000g/mL
Boiling Point 101°C to 102°C
Flash Point -6°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)3CCH2C(CH3)=CH2
Refractive Index 1.4070 to 1.409
Quantity 100 mL
Specific Gravity 0.7
Solubility Information Solubility in water: immiscible with water
Formula Weight 112.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 4, 4-Trimethyl-1-pentene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,4,4-Trimethyl-1-pentene
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : 2,4,4-Trimethyl-1-pentene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.