missing translation for 'onlineSavingsMsg'
Learn More

10-(1-Naphthyl)anthracene-9-boronic acid, 97%, Thermo Scientific Chemicals

Catalog Number 15459617
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
400607-46-7
C24H17BO2
348.21
MFCD11977302
ASQXKNXJNDLXQV-UHFFFAOYSA-N
10-naphthalen-1-yl anthracen-9-yl boronic acid, 10-1-naphthalenyl-9-anthracenyl boronic acid, 10-naphthalene-1-yl-9-anthracene boronic acid, 10-1-naphthyl anthracene-9-boronic acid, 10-naphthalen-1-yl anthracen-9-ylboronic acid, 10-napthalene-1-yl-9-anthracene boronic acid, 10-1-naphthyl-9-anthryl boronic acid, 10-1-naphthyl anthracene-9-ylboronic acid
23088558
(10-naphthalen-1-ylanthracen-9-yl)boronic acid
OB(O)C1=C2C=CC=CC2=C(C2=CC=CC=C12)C1=C2C=CC=CC2=CC=C1
This item is not returnable. View return policy
400607-46-7
C24H17BO2
348.21
MFCD11977302
ASQXKNXJNDLXQV-UHFFFAOYSA-N
10-naphthalen-1-yl anthracen-9-yl boronic acid, 10-1-naphthalenyl-9-anthracenyl boronic acid, 10-naphthalene-1-yl-9-anthracene boronic acid, 10-1-naphthyl anthracene-9-boronic acid, 10-naphthalen-1-yl anthracen-9-ylboronic acid, 10-napthalene-1-yl-9-anthracene boronic acid, 10-1-naphthyl-9-anthryl boronic acid, 10-1-naphthyl anthracene-9-ylboronic acid
23088558
(10-naphthalen-1-ylanthracen-9-yl)boronic acid
OB(O)C1=C2C=CC=CC2=C(C2=CC=CC=C12)C1=C2C=CC=CC2=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 348.21
Percent Purity 97%
Chemical Name or Material 10-(1-Naphthyl)anthracene-9-boronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'