missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-2-piperidnyl)methanamine, 97%, Thermo Scientific™

Product Code. p-4510162
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10612025 250 mg Amber Glass Bottle 250mg
10696483 1 g Amber Glass Bottle 1g
10450584 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10612025 Supplier Thermo Scientific Maybridge Supplier No. MO01154CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 5298-72-6
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.22
MDL Number MFCD05022432
InChI Key PPUMJZMVFCLQBI-UHFFFAOYNA-N
PubChem CID 2794677
SMILES CN1CCCCC1CN

Specifications

Density 0.9102g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 1-methylpiperidin-2-yl methanamine, 1-methyl-2-piperidinyl methanamine, 2-piperidinemethanamine, 1-methyl, 1-1-methylpiperidin-2-yl methanamine, c-1-methyl-piperidin-2-yl-methylamine, 1-methyl-2-piperidnyl methanamine, 1-methyl-2-piperidyl methylamine, pubchem8033, 1-methyl-2-aminomethylpiperidine, 2-piperidinemethanamine,1-methyl
IUPAC Name (1-methylpiperidin-2-yl)methanamine
Formula Weight 128.22
Percent Purity 97%
Chemical Name or Material (1-Methyl-2-piperidinyl)methanamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.